4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA108591000
Price (MYR)
271.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
271.00
Add to cart
Chemical Identifiers
CAS57-71-6
IUPAC Name(3E)-3-(hydroxyimino)butan-2-one
Molecular FormulaC4H7NO2
InChI KeyFSEUPUDHEBLWJY-HWKANZROSA-N
SMILESCC(=O)C(\C)=N\O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Sulfated ash=<0.1 %
Solubility(5 % in water) Clear to slightly hazy
Appearance (Color)White to off-white