4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number FSA108590050
Price (MYR)
284.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
284.00
Add to cart
Chemical Identifiers
CAS75-75-2
IUPAC Namemethanesulfonic acid
Molecular FormulaCH4O3S
InChI KeyAFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILESCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<150 APHA
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4285 to 1.4315 (20°C, 589 nm)