4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chloroaniline, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA108590010
Price (MYR)
1,006.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,006.00
Add to cart
Chemical Identifiers
CAS766-84-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light beige
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Crystals and/or chunks or fused solid
Melting point38°C to 42°C