2-chloroaniline, 98+%, Thermo Scientific Chemicals
2-chloroaniline, 98+%, Thermo Scientific Chemicals
2-chloroaniline, 98+%, Thermo Scientific Chemicals
2-chloroaniline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-chloroaniline, 98+%, Thermo Scientific Chemicals
CAS: 95-51-2 | C6H6ClN | 127.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA108572500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Chloroaniline
Melting Point-3°C
CAS95-51-2
CAS Min %98.0
ColorYellow to Brown