4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
CAS: 99-91-2 | C8H7ClO | 154.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
Glass bottle
Catalog number FSA108542500
Price (MYR)
490.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
490.00
Add to cart
Chemical Identifiers
CAS477-30-5
IUPAC Name3,4,5,14-tetramethoxy-10-(methylamino)tricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-13-one
Molecular FormulaC21H25NO5
InChI KeyNNJPGOLRFBJNIW-UHFFFAOYNA-N
SMILESCNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)Pale yellow
FormPowder
Proton NMRConforms