4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4'-Chloroacetophenone, 98+%, Thermo Scientific Chemicals
CAS: 99-91-2 | C8H7ClO | 154.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
Packaging:
Glass bottle
Catalog number FSA108540050
Price (MYR)
224.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
224.00
Add to cart
Chemical Identifiers
CAS1335-32-6
IUPAC Nametris(λ²-lead(2+)) diacetate tetrahydroxide
Molecular FormulaC4H10O8Pb3
InChI KeyVLOJXAQYHIVPFI-UHFFFAOYSA-H
SMILES[OH-].[OH-].[OH-].[OH-].[Pb++].[Pb++].[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material≤0.01%
Impurity contentChloride (Cl): ≤5 ppm
Impurity contentCopper (Cu): ≤0.002%
Impurity contentIron (Fe): ≤0.001%
Impurity contentNitrate and Nitrite (as NO₃): ≤0.005%