Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Chloroacetaldehyde dimethyl acetal, 97%, Thermo Scientific Chemicals
CAS: 97-97-2 | C4H9ClO2 | 124.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA108500010
Price (MYR)
564.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
564.00
Add to cart
Chemical Identifiers
CAS4347-33-5
IUPAC Name(5-formylthiophen-2-yl)boronic acid
Molecular FormulaC5H5BO3S
InChI KeyDEQOVKFWRPOPQP-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(S1)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to orange
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %