D(+)-Cellobiose, 98%
D(+)-Cellobiose, 98%
D(+)-Cellobiose, 98%
D(+)-Cellobiose, 98%
Thermo Scientific Chemicals
D(+)-Cellobiose, 98%
CAS: 528-50-7 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA108461000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialD(+)-Cellobiose
CAS528-50-7
Melting Point239°C
ColorWhite
EINECS Number208-436-5