Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
25 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA108262500
Price (MYR)
664.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
664.00
Add to cart
Chemical Identifiers
CAS9001-73-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light brown
Infrared spectrumConforms
Assay>=1000 TU/mg (Enz.act.) accurancy:+/-5%
Loss on drying=<7.0 % (60°C, 4 hrs) (vacuum)
pH4.8 to 6.2 (1 in 50 solution)