Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
250 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA108260250
Price (MYR)
320.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
320.00
Add to cart
Chemical Identifiers
CAS111-65-9
IUPAC Nameoctane
Molecular FormulaC8H18
InChI KeyTVMXDCGIABBOFY-UHFFFAOYSA-N
SMILESCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Density(15°C) 0.706 to 0.710 kg/l
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid