Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Carbazole, 96%, Thermo Scientific Chemicals
CAS: 86-74-8 | C12H9N | 167.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
25 g
250 g
Packaging:
View all
Plastic bottle
Catalog number FSA108260010
Price (MYR)
1,874.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,874.00
Add to cart
Chemical Identifiers
CAS1674-37-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=99.0 %
Infrared spectrumConforms
Refractive index1.5024 to 1.5054 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow