ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
ε-Caprolactam, 99+%, Thermo Scientific Chemicals
CAS: 105-60-2 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA108230025
Price (MYR)
687.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
687.00
Add to cart
Chemical Identifiers
CAS1310-65-2
IUPAC Namelithium(1+) hydroxide
Molecular FormulaHLiO
InChI KeyWMFOQBRAJBCJND-UHFFFAOYSA-M
SMILES[Li+].[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormGranules or pellets
Assay (special)?2.5 % of Theoretical value (Li)
Total Metal Impurities≤0.005% (50 ppm)