D(+)-10-Camphorsulfonic acid, 99%, Thermo Scientific Chemicals
D(+)-10-Camphorsulfonic acid, 99%, Thermo Scientific Chemicals
D(+)-10-Camphorsulfonic acid, 99%, Thermo Scientific Chemicals
D(+)-10-Camphorsulfonic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
D(+)-10-Camphorsulfonic acid, 99%, Thermo Scientific Chemicals
CAS: 3144-16-9 | C10H15O4S | 231.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA108221000
Price (MYR)
457.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
457.00
Add to cart
Specifications
Chemical Name or MaterialD(+)-10-Camphorsulfonic acid
Name Note99%
CAS3144-16-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement