Calmagite, Indicator Grade, Pure
Calmagite, Indicator Grade, Pure
Calmagite, Indicator Grade, Pure
Calmagite, Indicator Grade, Pure
Thermo Scientific Chemicals
Calmagite, Indicator Grade, Pure
CAS: 3147-14-6 | C17H14N2O5S | 358.368 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
10 g
25 g
Packaging:
Glass bottle
Catalog number FSA108171000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCalmagite
Melting Point300.0°C
CAS3147-14-6
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
View more
Indicator
RUO – Research Use Only