n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
CAS: 107-92-6 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA108110025
Price (MYR)
514.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
514.00
Add to cart
Chemical Identifiers
CAS3761-94-2
IUPAC Name1-methylcycloheptan-1-ol
Molecular FormulaC8H16O
InChI KeyXFFKAYOHINCUNU-UHFFFAOYSA-N
SMILESCC1(O)CCCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.4700-1.4740 @ 20?C
Assay (GC)≥97.5%