n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyric acid, +99%, Thermo Scientific Chemicals
CAS: 107-92-6 | C4H8O2 | 88.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA108110010
Price (MYR)
394.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
394.00
Add to cart
Specifications
Chemical Name or Materialn-Butyric acid
CAS107-92-6
Melting Point-7.0°C to +5.0°C
Boiling Point162.0°C to 165.0°C
CAS Min %99.0