n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Catalog number FSA108015000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS102-82-9
IUPAC Nametributylamine
Molecular FormulaC12H27N
InChI KeyIMFACGCPASFAPR-UHFFFAOYSA-N
SMILESCCCCN(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.2 %
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4273 to 1.4293 (20°C, 589 nm)
Appearance (Form)Clear liquid