n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl sulfide, 99%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA108011000
Price (MYR)
424.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
424.00
Add to cart
Specifications
Chemical Name or Materialn-Butyl sulfide
CAS544-40-1
Health Hazard 1GHS Signal Word: Warning
Melting Point-80.0°C
ColorColorless to Yellow