4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylphenol, 97%, Thermo Scientific Chemicals
CAS: 98-54-4 | C10H14O | 150.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 g
Catalog number FSA108000010
Price (MYR)
432.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
432.00
Add to cart
Chemical Identifiers
CAS10035-10-6
IUPAC Namehydrogen bromide
Molecular FormulaBrH
InChI KeyCPELXLSAUQHCOX-UHFFFAOYSA-N
SMILESBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
Titration with NaOH47.0 to 49.0 %
Heavy metals (ICP-OES)=<5 ppm
Residue after ignition=<0.002 %