n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl disulfide, 97%, Thermo Scientific Chemicals
CAS: 629-45-8 | C8H18S2 | 178.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA107920250
Price (MYR)
318.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
318.00
Add to cart
Chemical Identifiers
CAS6020-87-7
IUPAC Name2-(N-methylcarbamimidamido)acetic acid hydrate
Molecular FormulaC4H11N3O3
InChI KeyMEJYXFHCRXAUIL-UHFFFAOYSA-N
SMILESO.CN(CC(O)=O)C(N)=N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Residue after ignition=<1 %
HPLC>=98.5 % (on dry substance)
Heavy metals (as Pb)=<10 ppm