4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
CAS: 98-52-2 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA107902500
Price (MYR)
522.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
522.00
Add to cart
Chemical Identifiers
CAS536-90-3
IUPAC Name3-methoxyaniline
Molecular FormulaC7H9NO
InChI KeyNCBZRJODKRCREW-UHFFFAOYSA-N
SMILESCOC1=CC=CC(N)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5790-1.5830 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Pale yellow to dark red or dark brown