4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-tert-Butylcyclohexanol, 99%, mixture of isomers, Thermo Scientific Chemicals
CAS: 98-52-2 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
Packaging:
View all
Glass Bottle
Catalog number FSA107900050
Price (MYR)
203.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
203.00
Add to cart
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver white
FormWire, 0.6mm (0.024in) diameter, hardened.
Assay from Supplier's CofA≥99.95% (metals basis)