n-Butylbenzene, 99+%, Thermo Scientific Chemicals
n-Butylbenzene, 99+%, Thermo Scientific Chemicals
n-Butylbenzene, 99+%, Thermo Scientific Chemicals
n-Butylbenzene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butylbenzene, 99+%, Thermo Scientific Chemicals
CAS: 104-51-8 | C10H14 | 134.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA107852500
Price (MYR)
867.00
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
867.00
Add to cart
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Trace MetalSodium, Na + potassium, K: ≤0.2%
Appearance (Color)Clear colorless to yellow
Assay (Titration)≥52.5%
Residual halide≤0.5% as bromide