tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
CAS: 540-88-5 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass bottle
Catalog number FSA107760025
Price (MYR)
3,375.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,375.00
Add to cart
Chemical Identifiers
CAS1196-01-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4950 to 1.4970 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=92 %