sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic Drum
Catalog number FSA107700100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS631-61-8
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
CommentComponent is dissolved in 18.2 megohm-cm ± 1 water, followed by pH adjustment and 0.22 micron filtration
pH<8.0
Appearance (Color)Clear colorless