sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
sec-Butanol, 99%, Extra Pure, Thermo Scientific Chemicals
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
10 L
Packaging:
View all
Glass Bottle
Catalog number FSA107700025
Price (MYR)
598.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
598.00
Add to cart
Chemical Identifiers
CAS3167-49-5
IUPAC Name6-aminopyridine-3-carboxylic acid
Molecular FormulaC6H6N2O2
InChI KeyZCIFWRHIEBXBOY-UHFFFAOYSA-N
SMILESNC1=CC=C(C=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration with HClO4>=97.5 %
Appearance (Color)White to beige