1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
CAS: 1633-83-6 | C4H8O3S | 136.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA107671000
Price (MYR)
878.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
878.00
Add to cart
Chemical Identifiers
CAS552-16-9
IUPAC Name2-nitrobenzoic acid
Molecular FormulaC7H5NO4
InChI KeySLAMLWHELXOEJZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Melting point143°C to 149°C
Infrared spectrumConforms
HPLC>=94 %
Appearance (Form)Powder