1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Butane sultone, 99+%, Thermo Scientific Chemicals
CAS: 1633-83-6 | C4H8O3S | 136.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA107670250
Price (MYR)
434.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
434.00
Add to cart
Chemical Identifiers
CAS3033-62-3
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Refractive Index1.4280-1.4320 @ 20?C
Assay (GC)≥97.5%