2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,3-Butanedione, 99%, Thermo Scientific Chemicals
CAS: 431-03-8 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA107651000
Price (MYR)
596.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
596.00
Add to cart
Specifications
Chemical Name or Material2,3-Butanedione
Name Note99%
CAS431-03-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger