Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
CAS: 77-79-2 | C4H6O2S | 118.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA107615000
Price (MYR)
637.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
637.00
Add to cart
Chemical Identifiers
CAS108-38-3
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.005 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=99.0 %