Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butadiene sulfone, 98%, Thermo Scientific Chemicals
CAS: 77-79-2 | C4H6O2S | 118.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA107611000
Price (MYR)
259.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
259.00
Add to cart
Chemical Identifiers
CAS35963-20-3
IUPAC Name[(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
Molecular FormulaC10H16O4S
InChI KeyMIOPJNTWMNEORI-MHPPCMCBSA-N
SMILESCC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Optical Rotation-21.0? ? 2.5? (c=20 in water)
Appearance (Color)White to cream