N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
10 g
100 g
1 kg
Packaging:
View all
Plastic drum
Catalog number FSA107450050
Price (MYR)
4,006.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
4,006.00
Add to cart
Chemical Identifiers
CAS100-27-6
IUPAC Name2-(4-nitrophenyl)ethan-1-ol
Molecular FormulaC8H9NO3
InChI KeyIKMXRUOZUUKSON-UHFFFAOYSA-N
SMILESOCCC1=CC=C(C=C1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥97.5%
FormPowder and/or Lumps
Appearance (Color)Yellow to yellow-orange to brown
Coupling reagent, C-X bond formation reagent
RUO – Research Use Only