N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Bromosuccinimide, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
10 g
100 g
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA107450010
Price (MYR)
1,120.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,120.00
Add to cart
Chemical Identifiers
CAS4835-90-9
IUPAC Name3-hydroxy-2,2-dimethylpropanoic acid
Molecular FormulaC5H10O3
InChI KeyRDFQSFOGKVZWKF-UHFFFAOYSA-N
SMILESCC(C)(CO)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Water Content (Karl Fischer Titration)≤1%
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
View more
Coupling reagent, C-X bond formation reagent
RUO – Research Use Only