3-Bromopyridine, 99%, Thermo Scientific Chemicals
3-Bromopyridine, 99%, Thermo Scientific Chemicals
3-Bromopyridine, 99%, Thermo Scientific Chemicals
3-Bromopyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Bromopyridine, 99%, Thermo Scientific Chemicals
CAS: 626-55-1 | C5H4BrN | 158.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA107410250
Price (MYR)
483.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
483.00
Add to cart
Specifications
Chemical Name or Material3-Bromopyridine
Name Note99%
CAS626-55-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger