1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromo-3-phenylpropane, 98%, Thermo Scientific Chemicals
CAS: 637-59-2 | C9H11Br | 199.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA107295000
Price (MYR)
3,185.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,185.00
Add to cart
Chemical Identifiers
CAS150884-56-3
IUPAC Nametert-butyl (3S)-3-(4-cyanophenyl)oxaziridine-2-carboxylate
Molecular FormulaC13H14N2O3
InChI KeyACXPNVRTMHEHMQ-VPHXOMNUSA-N
SMILESCC(C)(C)OC(=O)N1O[C@H]1C1=CC=C(C=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point57°C to 63°C
Elemental Analysis>=98.0 %