1-Bromononane, 97%, Thermo Scientific Chemicals
1-Bromononane, 97%, Thermo Scientific Chemicals
1-Bromononane, 97%, Thermo Scientific Chemicals
1-Bromononane, 97%, Thermo Scientific Chemicals
1-Bromononane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromononane, 97%, Thermo Scientific Chemicals
CAS: 693-58-3 | C9H19Br | 207.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA107205000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS603-34-9
IUPAC NameN,N-diphenylaniline
Molecular FormulaC18H15N
InChI KeyODHXBMXNKOYIBV-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light beige
Melting point125°C to 127°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=99.0 %
Suitable for battery materials development.
RUO – Research Use Only