5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Bromoindole, 99%, Thermo Scientific Chemicals
CAS: 10075-50-0 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA107060250
Price (MYR)
658.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
658.00
Add to cart
Chemical Identifiers
CAS95-55-6
IUPAC Name2-aminophenol
Molecular FormulaC6H7NO
InChI KeyCDAWCLOXVUBKRW-UHFFFAOYSA-N
SMILESNC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Melting Point (clear melt)172-177?C
Appearance (Color)White to grey to brown
Assay (GC)≥98.5%