1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
1 L
Packaging:
Glass bottle
Catalog number FSA107032500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2386-52-9
IUPAC Namemethanesulfonic acid silver
Molecular FormulaCH4AgO3S
InChI KeyGTBQRHOYAUGRPV-UHFFFAOYSA-N
SMILES[Ag].CS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)50% Ag min