1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA107030050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-Bromoheptane
Name Note99%
CAS629-04-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning