1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Bromoheptane, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
5 mL
250 mL
Packaging:
Glass bottle
Catalog number FSA107030010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7718-98-1
IUPAC Namevanadium(3+) trichloride
Molecular FormulaCl3V
InChI KeyHQYCOEXWFMFWLR-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[V+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=96.0 %
Appearance (Form)Powder
Insoluble matter=<0.5 % (in water)
Appearance (Color)Dark blue to violet to black