4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Bromofluorobenzene, 99%, Thermo Scientific Chemicals
CAS: 460-00-4 | C6H4BrF | 175.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
Packaging:
Glass bottle
Catalog number FSA107010100
Price (MYR)
261.00
Add to cart
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
261.00
Add to cart
Specifications
Chemical Name or Material4-Bromofluorobenzene
Name Note99%
CAS460-00-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning