2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoethylamine hydrobromide, 99%, Thermo Scientific Chemicals
CAS: 2576-47-8 | C2H6BrN·HBr | 204.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
250 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA106931000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Bromoethylamine hydrobromide
Name Note99%
CAS2576-47-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning