2-Bromobenzonitrile, 99%
2-Bromobenzonitrile, 99%
2-Bromobenzonitrile, 99%
2-Bromobenzonitrile, 99%
Thermo Scientific Chemicals
2-Bromobenzonitrile, 99%
CAS: 2042-37-7 | C7H4BrN | 182.02 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA10669010010 gGlass bottle
Catalog number FSA106690100
Price (MYR)
666.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
666.00
Add to cart
Chemical Identifiers
CAS119397-06-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%