Biuret, 97%, extra pure
Biuret, 97%, extra pure
Biuret, 97%, extra pure
Biuret, 97%, extra pure
Thermo Scientific Chemicals
Biuret, 97%, extra pure
CAS: 108-19-0 | C2H5N3O2 | 103.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA106520250
Price (MYR)
508.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
508.00
Add to cart
Chemical Identifiers
CAS1195-45-5
IUPAC Name1-fluoro-4-isocyanatobenzene
Molecular FormulaC7H4FNO
InChI KeyDSVGFKBFFICWLZ-UHFFFAOYSA-N
SMILESFC1=CC=C(C=C1)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.5130 to 1.5150 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow