Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA106230250
Price (MYR)
311.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
311.00
Add to cart
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.025mm (0.001in) diameter, hardened.
Assay from Supplier's CofA≥99.98% (metals basis)
Appearance (Color)Silvery white