Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bibenzyl, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number FSA106230050
Price (MYR)
225.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
225.00
Add to cart
Specifications
Chemical Name or MaterialBibenzyl
Name Note99%
CAS103-29-7
CAS Min %98.5
CAS Max %100.0