Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzylamine, 99%, pure, Thermo Scientific Chemicals
CAS: 100-46-9 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA105851000
Price (MYR)
246.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
246.00
Add to cart
Chemical Identifiers
CAS873-31-4
IUPAC Name1-chloro-2-ethynylbenzene
Molecular FormulaC8H5Cl
InChI KeyDGLHLIWXYSGYBI-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)97.5% min