Benzoyl chloride, 99.5+%, pure, Thermo Scientific Chemicals
Benzoyl chloride, 99.5+%, pure, Thermo Scientific Chemicals
Benzoyl chloride, 99.5+%, pure, Thermo Scientific Chemicals
Benzoyl chloride, 99.5+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoyl chloride, 99.5+%, pure, Thermo Scientific Chemicals
Benzoyl Chloride, 99%, C7H5ClO, CAS Number-98-88-4, unii-vty8706w36, a-chlorobenzaldehyde, benzaldehyde, alpha-chloro, benzoic acid chloride, benzenecarbonyl chloride, benzoic acid, chloride, alpha-chlorobenzaldehyde, benzoylchloride, ccris 802, benzoylchlorid, 2.5L, 98.5% min. (GC), 09, 182
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA105750025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS10042-76-9
IUPAC Namestrontium(2+) dinitrate
Molecular FormulaN2O6Sr
InChI KeyDHEQXMRUPNDRPG-UHFFFAOYSA-N
SMILES[Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=97.5 %
Loss on drying=<0.5 %
pH5 to 7 (5 % at 20°C)
Heavy metals (as Pb)=<10 ppm
Appearance (Form)Crystalline powder or crystals
View more
Benzoyl Chloride, 99%, C7H5ClO, CAS Number-98-88-4, unii-vty8706w36, a-chlorobenzaldehyde, benzaldehyde, alpha-chloro, benzoic acid chloride, benzenecarbonyl chloride, benzoic acid, chloride, alpha-chlorobenzaldehyde, benzoylchloride, ccris 802, benzoylchlorid, 2.5L, 98.5% min. (GC), 09, 182
RUO – Research Use Only