Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
5 mg
25 mg
Packaging:
Glass bottle
Catalog number FSA105551000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS593-56-6
IUPAC Namehydrogen O-methylhydroxylamine chloride
Molecular FormulaCH6ClNO
InChI KeyXNXVOSBNFZWHBV-UHFFFAOYSA-N
SMILES[H+].[Cl-].CON
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)White to light yellow
Melting point148°C to 153°C
Titration with HClO498.0 to 102.0 %