Benzoin, 98%, Thermo Scientific Chemicals
Benzoin, 98%, Thermo Scientific Chemicals
Benzoin, 98%, Thermo Scientific Chemicals
Benzoin, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzoin, 98%, Thermo Scientific Chemicals
CAS: 119-53-9 | C14H12O2 | 212.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA105511000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzoin
Name Note98%
CAS119-53-9
CAS Min %97.5
CAS Max %100.0